Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099286.2 | NP_001092756.1 | 6 | 1175 | ||
NM_001318738.1 | NP_001305667.1 | 7 | 987 | |||
NM_138419.4 | NP_612428.2 | 6 | 887 | |||
XM_011535410.2 | XP_011533712.1 | 7 | 1239 | |||
XM_011535412.2 | XP_011533714.1 | 5 | 980 | |||
XM_011535413.2 | XP_011533715.1 | 5 | 4094 | |||
GenBank mRNA | AK125758.1 | - | 7 | 944 | ||
AK292303.1 | - | 6 | 1126 | |||
BC011716.2 | - | 3 | 510 | |||
BC063688.1 | - | 6 | 840 | 4455541 |