Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138421.2 | NP_612430.2 | 6 | 596 | ||
GenBank mRNA | AB489136.1 | - | 6 | 546 | ||
AK123457.1 | - | 6 | 590 | |||
BC012010.1 | - | 6 | 578 | 4473003 | ||
BU150462.1 | - | 2 | 167 | |||
DR155913.1 | - | 6 | 596 |