Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2U1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183075.2 | NP_898898.1 | 4 | 1468 | ||
XM_005262717.2 | XP_005262774.1 | 4 | 1534 | |||
XM_005262720.2 | XP_005262777.1 | 3 | 844 | |||
GenBank mRNA | AK290002.1 | - | 4 | 1473 | ||
AK298989.1 | - | 3 | 390 | |||
AK303284.1 | - | 5 | 1273 | |||
AK304724.1 | - | 5 | 1346 | |||
AY343323.1 | - | 4 | 1424 | |||
BC132767.1 | - | 4 | 1433 | 40146789 | ||
BC136483.1 | - | 4 | 1433 | 9020472 |