Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286136.1 | NP_001273065.1 | 2, 3 | 403 | ||
NM_053280.4 | NP_444510.2 | 2, 3 | 403 | |||
GenBank mRNA | AB067774.1 | - | 2, 3 | 510 | ||
AL133658.1 | - | 1, 2 | 622 | |||
AY713299.1 | - | 2, 3 | 378 | |||
BC126222.1 | - | 2, 3 | 211 | 8991938 | ||
BC126248.1 | - | 2, 3 | 211 | 8991964 | ||
BC144022.1 | - | 2, 3 | 211 | 9052542 | ||
FJ436414.1 | - | 1, 2 | 111 |