Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005222.3 | NP_005213.3 | 3 | 689 | ||
GenBank mRNA | AK094086.1 | - | 3 | 500 | ||
BC069363.1 | - | 3 | 335 | 7262091 | ||
BC103688.1 | - | 3 | 368 | 40000523 | ||
BC103689.1 | - | 3 | 368 | 40000521 | ||
BC103690.1 | - | 3 | 368 | 40000520 | ||
BC109381.1 | - | 3 | 368 | 40000518 |