Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172420.1 | NP_001165891.1 | 7 | 1189 | ||
NM_018100.3 | NP_060570.2 | 6 | 1277 | |||
NR_033327.1 | 5 | 2534 | ||||
GenBank mRNA | AK001328.1 | - | 6 | 1163 | ||
AK225543.1 | - | 5 | 2426 | |||
AK295064.1 | - | 6 | 1020 | |||
AK297632.1 | - | 7 | 1189 | |||
AL122084.1 | - | 5 | 2420 | |||
BC020210.1 | - | 6 | 1153 | 4586338 | ||
EU520261.1 | - | 6 | 1162 |