Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076780.1 | NP_001070248.1 | 16 | 2755 | ||
NM_001278423.1 | NP_001265352.1 | 5 | 849 | |||
NM_001278425.1 | NP_001265354.1 | 5 | 849 | |||
NM_052892.3 | NP_443124.3 | 16 | 2755 | |||
NR_126532.1 | 16 | 2755 | ||||
GenBank mRNA | AB067466.1 | - | 8 | 1100 | ||
AK098052.1 | - | 10 | 1337 | |||
AK127266.1 | - | 4 | 637 | |||
AY164483.1 | - | 16 | 2755 | |||
AY371495.1 | - | 5 | 811 | |||
BC004562.2 | - | 5 | 816 | |||
BC014157.2 | - | 5 | 816 | |||
CR627341.1 | - | 3 | 342 |