Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099272.1 | NP_001092742.1 | 8 | 1779 | ||
NM_001172418.1 | NP_001165889.1 | 8 | 1651 | |||
NM_052893.1 | NP_443125.1 | 9 | 1979 | |||
NM_152733.2 | NP_689946.2 | 7 | 1516 | |||
XM_011514279.2 | XP_011512581.1 | 10 | 2376 | |||
GenBank mRNA | AB067467.1 | - | 8 | 1645 | ||
AK057507.1 | - | 7 | 1516 | |||
AK090930.1 | - | 4 | 483 | |||
BC101354.2 | - | 7 | 1437 | 40025568 | ||
BC101355.1 | - | 7 | 1437 | 40025569 | ||
BC101356.2 | - | 8 | 1506 | |||
BC101357.1 | - | 8 | 1572 | 40025572 | ||
BX537534.1 | - | 8 | 1510 |