Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281956.1 | NP_001268885.1 | 17, 18 | 2867 | ||
NM_052896.4 | NP_443128.2 | 17, 18 | 2599 | |||
XM_017000185.1 | XP_016855674.1 | 17, 18 | 2861 | |||
XM_017000186.1 | XP_016855675.1 | 17, 18 | 2861 | |||
XM_017000187.1 | XP_016855676.1 | 17, 18 | 2690 | |||
XM_017000188.1 | XP_016855677.1 | 17, 18 | 2599 | |||
XM_017000189.1 | XP_016855678.1 | 17, 18 | 2861 | |||
XM_017000190.1 | XP_016855679.1 | 11, 12 | 2126 | |||
XM_017000191.1 | XP_016855680.1 | 17, 18 | 2861 | |||
XM_017000192.1 | XP_016855681.1 | 17, 18 | 2861 | |||
XM_017000193.1 | XP_016855682.1 | 17, 18 | 2861 | |||
XM_017000194.1 | XP_016855683.1 | 17, 18 | 2599 | |||
GenBank mRNA | AB212622.1 | - | 17, 18 | 2867 | ||
AY210418.1 | - | 17, 18 | 2599 |