Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052900.2 | NP_443132.3 | 23 | 3838 | ||
NM_198123.1 | NP_937756.1 | 24 | 4150 | |||
NM_198124.1 | NP_937757.1 | 25 | 4042 | |||
XM_011516816.2 | XP_011515118.1 | 23 | 4324 | |||
XM_017013008.1 | XP_016868497.1 | 23 | 4558 | |||
XM_017013009.1 | XP_016868498.1 | 24 | 4636 | |||
XM_017013010.1 | XP_016868499.1 | 22 | 4246 | |||
XM_017013011.1 | XP_016868500.1 | 22 | 4246 | |||
XM_017013012.1 | XP_016868501.1 | 22 | 3609 | |||
GenBank mRNA | AB067481.2 | - | 11 | 2011 | ||
AB114604.1 | - | 24 | 4150 | |||
AB114605.1 | - | 25 | 4042 | |||
AY210419.1 | - | 23 | 3709 |