Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006641.3 | NP_001006642.1 | 2 | 493 | ||
NM_001006642.3 | NP_001006643.1 | 2 | 738 | |||
NM_001265614.2 | NP_001252543.1 | 2 | 738 | |||
NM_052901.4 | NP_443133.2 | 2 | 356 | |||
NR_049766.2 | 2 | 399 | ||||
XM_005251688.1 | XP_005251745.1 | 2 | 493 | |||
XM_005251689.4 | XP_005251746.1 | 2 | 752 | |||
XM_006716948.2 | XP_006717011.1 | 2 | 894 | |||
XM_006716949.3 | XP_006717012.1 | 2 | 754 | |||
GenBank mRNA | AB067483.1 | - | 2 | 737 | ||
AJ619963.1 | - | 2 | 657 | |||
AJ619989.1 | - | 2 | 281 | |||
AJ619990.1 | - | 2 | 376 | |||
AJ619991.1 | - | 2 | 338 | |||
AJ619992.1 | - | 2 | 324 | |||
AK290705.1 | - | 2 | 329 | |||
AK290991.1 | - | 2 | 493 | |||
AL559911.3 | - | 2 | 500 | |||
BC089448.1 | - | 2 | 347 | 30558551 | ||
BC103930.1 | - | 2 | 298 | 40008034 | ||
BC103931.1 | - | 2 | 298 | 40008035 | ||
BC103932.1 | - | 2 | 298 | 40008036 | ||
BC103933.1 | - | 2 | 298 | 40008037 | ||
BQ637100.1 | - | 2 | 356 |