Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052905.3 | NP_443137.2 | 23 | 3302 | ||
XM_005246263.2 | XP_005246320.1 | 23 | 3369 | |||
XM_005246265.3 | XP_005246322.1 | 23 | 3369 | |||
XM_011510530.1 | XP_011508832.1 | 24 | 3390 | |||
XM_011510531.1 | XP_011508833.1 | 24 | 3390 | |||
XM_011510532.2 | XP_011508834.1 | 24 | 3390 | |||
XM_011510533.2 | XP_011508835.1 | 24 | 3390 | |||
XM_011510534.1 | XP_011508836.1 | 23 | 3321 | |||
XM_011510535.2 | XP_011508837.1 | 23 | 3237 | |||
XM_011510536.2 | XP_011508838.1 | 18 | 2341 | |||
GenBank mRNA | AB067489.1 | - | 23 | 3073 | ||
AK094865.1 | - | 7 | 857 | |||
AK131316.1 | - | 9 | 1500 | |||
AL834396.1 | - | 21 | 2686 | |||
BC113878.1 | - | 10 | 1264 | |||
BC114438.1 | - | 10 | 1269 | |||
BC167159.1 | - | 23 | 3073 | 8862581 | ||
BQ067763.1 | - | 1 | 59 |