Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081560.2 | NP_001075029.1 | 4 | 615 | ||
NM_001081562.2 | NP_001075031.1 | 4 | 615 | |||
NM_001081563.2 | NP_001075032.1 | 3 | 984 | |||
NM_001288764.1 | NP_001275693.1 | 5 | 674 | |||
NM_001288765.1 | NP_001275694.1 | 2 | 255 | |||
NM_001288766.1 | NP_001275695.1 | 4 | 615 | |||
NM_004409.4 | NP_004400.4 | 4 | 615 | |||
GenBank mRNA | AB209363.1 | - | 4 | 1458 | ||
AK297299.1 | - | 2 | 206 | |||
AK302874.1 | - | 4 | 548 | |||
AK304749.1 | - | 3 | 887 | |||
BC026328.1 | - | 5 | 669 | 4792104 | ||
BC062553.1 | - | 4 | 536 | 6480902 | ||
DC420602.1 | - | 2 | 238 | |||
KJ534826.1 | - | 3 | 440 | |||
KJ534827.1 | - | 3 | 440 | |||
KJ534828.1 | - | 3 | 440 | |||
L19268.1 | - | 4 | 537 | |||
M87312.1 | - | 3 | 985 | |||
M94203.1 | - | 4 | 287 | |||
S72883.1 | - | 4 | 446 |