Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FHAD1 SIRNA Assays ›
Gene Symbol:
FHAD1
Gene Name:
forkhead associated phosphopeptide binding domain 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 15236559 - 15400283 on Build GRCh38
UniGene ID:
Hs.659997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41675
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FHAD1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_052929.1
NP_443161.1
6
860
XM_011540575.2
XP_011538877.1
6
936
XM_011540576.2
XP_011538878.1
6
937
XM_011540577.2
XP_011538879.1
6
898
XM_011540580.2
XP_011538882.1
6
935
XM_011540581.2
XP_011538883.1
6
935
XM_011540582.2
XP_011538884.1
6
935
XM_011540584.2
XP_011538886.1
6
936
XM_011540588.2
XP_011538890.1
6
927
XM_011540589.2
XP_011538891.1
6
936
XM_011540590.2
XP_011538892.1
6
928
XM_011540592.2
XP_011538894.1
5
637
XM_011540593.2
XP_011538895.1
6
932
XM_011540595.2
XP_011538897.1
3
280
XM_011540597.2
XP_011538899.1
6
937
XM_011540598.2
XP_011538900.1
6
939
XM_017000198.1
XP_016855687.1
6
937
XM_017000199.1
XP_016855688.1
6
937
XM_017000200.1
XP_016855689.1
6
936
XM_017000201.1
XP_016855690.1
6
936
XM_017000202.1
XP_016855691.1
6
937
XM_017000203.1
XP_016855692.1
6
933
XM_017000204.1
XP_016855693.1
6
932
XM_017000205.1
XP_016855694.1
6
937
XM_017000206.1
XP_016855695.1
6
932
XM_017000208.1
XP_016855697.1
6
927
XM_017000209.1
XP_016855698.1
6
931
XM_017000210.1
XP_016855699.1
6
927
XM_017000211.1
XP_016855700.1
6
935
XM_017000212.1
XP_016855701.1
6
926
XM_017000213.1
XP_016855702.1
6
936
XM_017000214.1
XP_016855703.1
6
937
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0