Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OSBPL6 SIRNA Assays ›
Gene Symbol:
OSBPL6
Gene Name:
oxysterol binding protein like 6
Gene Aliases:
ORP6
Chromosome Location:
Chr.2: 178194479 - 178399433 on Build GRCh38
UniGene ID:
Hs.318775
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41686
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OSBPL6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001201480.1
NP_001188409.1
13
1631
NM_001201481.1
NP_001188410.1
11
1463
NM_001201482.1
NP_001188411.1
12
1556
NM_032523.3
NP_115912.1
12
1556
NM_145739.2
NP_665682.1
11
1144
XM_011510539.1
XP_011508841.1
12
1534
XM_017003265.1
XP_016858754.1
14
1703
XM_017003266.1
XP_016858755.1
14
1703
XM_017003267.1
XP_016858756.1
12
1398
XM_017003268.1
XP_016858757.1
13
1627
XM_017003269.1
XP_016858758.1
12
1398
XM_017003270.1
XP_016858759.1
12
1617
XM_017003271.1
XP_016858760.1
11
1323
XM_017003272.1
XP_016858761.1
12
1534
XM_017003273.1
XP_016858762.1
10
1366
XM_017003274.1
XP_016858763.1
11
1459
XM_017003275.1
XP_016858764.1
12
1553
XM_017003276.1
XP_016858765.1
11
1459
XM_017003277.1
XP_016858766.1
10
1366
GenBank mRNA
AB208898.1
-
13
1631
AF323728.1
-
12
1390
AF392448.1
-
12
1390
AF462443.1
-
11
1366
AK027600.1
-
5
431
AK093902.1
-
11
1146
AK300389.1
-
11
1421
BC021248.2
-
4
368
BC052259.1
-
11
1444
6428611
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
lipid transport
protein binding
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0