Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135674.1 | NP_001129146.1 | 3 | 997 | ||
NM_001135675.1 | NP_001129147.1 | 3 | 605 | |||
NM_001135676.1 | NP_001129148.1 | 3 | 290 | |||
NM_138436.3 | NP_612445.2 | 3 | 376 | |||
XM_005273398.4 | XP_005273455.1 | 3 | 911 | |||
GenBank mRNA | AK311932.1 | - | 3 | 351 | ||
BC013035.2 | - | 3 | 338 | 3535608 | ||
BI463786.1 | - | 3 | 295 | |||
BI549696.1 | - | 3 | 610 | |||
DA702519.1 | - | 3 | 376 |