Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001369.2 | NP_001360.1 | 79 | 13824 | ||
XM_005248262.3 | XP_005248319.2 | 79 | 13907 | |||
XM_017009177.1 | XP_016864666.1 | 77 | 13487 | |||
XM_017009178.1 | XP_016864667.1 | 79 | 14710 | |||
XM_017009179.1 | XP_016864668.1 | 73 | 13004 | |||
XM_017009185.1 | XP_016864674.1 | 51 | 9203 | |||
XM_017009186.1 | XP_016864675.1 | 48 | 8619 | |||
XM_017009188.1 | XP_016864677.1 | 45 | 9435 | |||
GenBank mRNA | AB046823.2 | - | 27 | 4888 | ||
AB290164.1 | - | 32 | 5827 | |||
AK026756.1 | - | 7 | 1180 | |||
AK128600.1 | - | 21 | 3827 | |||
AY045575.1 | - | 79 | 13824 |