Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FCRL1 SIRNA Assays ›
Gene Symbol:
FCRL1
Gene Name:
Fc receptor like 1
Gene Aliases:
CD307a, FCRH1, IFGP1, IRTA5
Chromosome Location:
Chr.1: 157792249 - 157820150 on Build GRCh38
UniGene ID:
Hs.656112
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41796
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FCRL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159397.1
NP_001152869.1
7
1097
NM_001159398.1
NP_001152870.1
8
1214
NM_052938.4
NP_443170.1
8
1214
XM_005244866.2
XP_005244923.1
8
1185
XM_005244867.2
XP_005244924.1
7
1068
XM_005244869.3
XP_005244926.1
6
910
XM_011509125.1
XP_011507427.1
8
1185
XM_011509126.1
XP_011507428.1
8
1185
XM_011509127.2
XP_011507429.1
7
1177
XM_011509128.1
XP_011507430.1
8
1185
XM_011509129.1
XP_011507431.1
8
1185
XM_011509130.1
XP_011507432.1
8
1185
XM_011509131.1
XP_011507433.1
8
1185
XM_011509132.2
XP_011507434.1
8
1185
XM_011509133.2
XP_011507435.1
8
1039
XM_011509134.2
XP_011507436.1
7
1014
XM_011509135.2
XP_011507437.1
7
931
XM_011509137.2
XP_011507439.1
5
559
XM_017000227.1
XP_016855716.1
7
3181
GenBank mRNA
AF329488.1
-
8
1124
AF459634.1
-
8
1167
AK096690.1
-
7
1036
AK315748.1
-
8
1142
AL833674.1
-
4
268
AL833970.1
-
4
1205
AY043464.1
-
8
1169
BC021274.2
-
8
882
BC033690.1
-
8
1165
5211491
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0