Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FCRL1 SIRNA Assays ›
Gene Symbol:
FCRL1
Gene Name:
Fc receptor like 1
Gene Aliases:
CD307a, FCRH1, IFGP1, IRTA5
Chromosome Location:
Chr.1: 157792249 - 157820150 on Build GRCh38
UniGene ID:
Hs.656112
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s41798
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FCRL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001159397.1
NP_001152869.1
9, 10
1203
NM_001159398.1
NP_001152870.1
10, 11
1327
NM_052938.4
NP_443170.1
10, 11
1330
XM_005244866.2
XP_005244923.1
10, 11
1291
XM_005244867.2
XP_005244924.1
9, 10
1184
XM_005244869.3
XP_005244926.1
8, 9
1026
XM_011509125.1
XP_011507427.1
11, 12
1299
XM_011509126.1
XP_011507428.1
11, 12
1296
XM_011509127.2
XP_011507429.1
9, 10
1293
XM_011509128.1
XP_011507430.1
10, 11
1303
XM_011509129.1
XP_011507431.1
10, 11
1300
XM_011509130.1
XP_011507432.1
10, 11
1288
XM_011509131.1
XP_011507433.1
11, 12
1311
XM_011509132.2
XP_011507434.1
11, 12
1308
XM_011509133.2
XP_011507435.1
10, 11
1155
XM_011509134.2
XP_011507436.1
9, 10
1130
XM_011509135.2
XP_011507437.1
9, 10
1047
XM_011509137.2
XP_011507439.1
7, 8
675
XM_017000227.1
XP_016855716.1
9, 10
3297
GenBank mRNA
AF329488.1
-
10, 11
1237
AF459634.1
-
10, 11
1283
AK096690.1
-
9, 10
1142
AK315748.1
-
10, 11
1258
AL833674.1
-
6, 7
374
AL833970.1
-
6, 7
1318
AY043464.1
-
10, 11
1285
BC021274.2
-
10, 11
998
BC033690.1
-
10, 11
1281
5211491
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0