Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138447.2 | NP_612456.1 | 3 | 856 | ||
NR_073480.1 | 3 | 491 | ||||
NR_073481.1 | 3 | 494 | ||||
NR_073482.1 | 3 | 374 | ||||
NR_104166.1 | 3 | 393 | ||||
GenBank mRNA | AB065282.1 | - | 3 | 844 | ||
AK074896.1 | - | 3 | 579 | |||
AK310459.1 | - | 3 | 413 | |||
BC014000.2 | - | 3 | 831 | 3936339 | ||
CD243730.1 | - | 1 | 41 | |||
CN406120.1 | - | 3 | 505 | |||
DA010230.1 | - | 3 | 495 | |||
DA485303.1 | - | 3 | 491 | |||
DB456506.1 | - | 4 | 374 | |||
HY005770.1 | - | 3 | 388 |