Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136050.2 | NP_001129522.1 | 9 | 1102 | ||
NM_138452.2 | NP_612461.1 | 9 | 1094 | |||
GenBank mRNA | AF418205.1 | - | 9 | 1038 | ||
AK058159.1 | - | 9 | 1094 | |||
AK126383.1 | - | 4 | 2287 | |||
AL050217.1 | - | 2 | 735 | |||
AL833917.1 | - | 9 | 997 | |||
BC014057.2 | - | 9 | 1008 | 3622913 | ||
BC015943.1 | - | 9 | 905 | 3922816 | ||
BX247980.1 | - | 9 | 1018 |