Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DYNC1LI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286157.1 | NP_001273086.1 | 6 | 648 | ||
NM_001323955.1 | NP_001310884.1 | 6 | 968 | |||
NM_006141.2 | NP_006132.1 | 7 | 879 | |||
NR_136658.1 | 7 | 1085 | ||||
XM_017023008.1 | XP_016878497.1 | 7 | 878 | |||
GenBank mRNA | AF035812.1 | - | 7 | 854 | ||
AK095237.1 | - | 1 | 1127 | |||
AK291766.1 | - | 7 | 858 | |||
AK295049.1 | - | 7 | 878 | |||
AK303031.1 | - | 6 | 627 | |||
AK304233.1 | - | 6 | 734 | |||
BC025959.1 | - | 7 | 854 | 4329417 | ||
EB387189.1 | - | 4 | 450 |