Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053282.4 | NP_444512.2 | 1, 2 | 258 | ||
GenBank mRNA | AF256653.1 | - | 1, 2 | 136 | ||
AF403479.1 | - | 1, 2 | 136 | |||
AF484964.1 | - | 1, 2 | 234 | |||
AK097162.1 | - | 1, 2 | 185 | |||
AK314743.1 | - | 1, 2 | 205 | |||
AL709120.1 | - | 1, 2 | 258 | |||
BC022407.1 | - | 1, 2 | 246 | 4717041 | ||
BC066595.1 | - | 1, 2 | 232 | 5755414 |