Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALNT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319051.1 | NP_001305980.1 | 4 | 1767 | ||
NM_054110.4 | NP_473451.3 | 4 | 1412 | |||
XM_005264852.4 | XP_005264909.1 | 4 | 944 | |||
XM_017005682.1 | XP_016861171.1 | 4 | 8825 | |||
XM_017005683.1 | XP_016861172.1 | 4 | 975 | |||
XM_017005684.1 | XP_016861173.1 | 4 | 975 | |||
GenBank mRNA | AB078149.1 | - | 4 | 937 | ||
AK094780.1 | - | 7 | 985 | |||
AK312425.1 | - | 4 | 1413 | |||
AL831925.1 | - | 4 | 1440 | |||
AL832575.2 | - | 4 | 914 | |||
AL832736.1 | - | 4 | 1382 | |||
AY035399.1 | - | 4 | 1220 | |||
AY358443.1 | - | 4 | 1398 | |||
BC014789.1 | - | 4 | 1362 | 4212702 |