Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311197.1 | NP_001298126.1 | 1 | 216 | ||
NM_080864.3 | NP_543140.1 | 1 | 216 | |||
GenBank mRNA | AB076563.1 | - | 1 | 159 | ||
AF447451.1 | - | 1 | 159 | |||
AY358181.1 | - | 2 | 999 | |||
BC110069.1 | - | 1 | 154 | |||
BC140935.1 | - | 1 | 216 | 9056747 |