Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001311197.1 | NP_001298126.1 | 3 | 371 | ||
NM_080864.3 | NP_543140.1 | 2 | 259 | |||
GenBank mRNA | AB076563.1 | - | 2 | 202 | ||
AF447451.1 | - | 2 | 202 | |||
AY358181.1 | - | 3 | 1042 | |||
BC110069.1 | - | 3 | 309 | |||
BC140935.1 | - | 2 | 259 | 9056747 |