Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207055.1 | NP_001193984.1 | 12 | 1560 | ||
NM_001207056.1 | NP_001193985.1 | 11 | 1458 | |||
NM_006892.3 | NP_008823.1 | 14 | 1746 | |||
NM_175848.1 | NP_787044.1 | 13 | 1686 | |||
NM_175849.1 | NP_787045.1 | 13 | 1686 | |||
NM_175850.2 | NP_787046.1 | 13 | 1665 | |||
XM_011528653.2 | XP_011526955.1 | 13 | 1538 | |||
XM_011528654.2 | XP_011526956.1 | 12 | 1413 | |||
GenBank mRNA | AB208880.1 | - | 13 | 1910 | ||
AF156487.1 | - | 13 | 1479 | |||
AF156488.1 | - | 14 | 1539 | |||
AF176228.1 | - | 13 | 1665 | |||
AF331857.1 | - | 14 | 1746 | |||
AK001191.1 | - | 7 | 618 | |||
AK295176.1 | - | 11 | 1160 | |||
AK299776.1 | - | 12 | 1393 | |||
AK299821.1 | - | 11 | 1201 | |||
AK299915.1 | - | 12 | 1337 | |||
DQ321787.1 | - | 13 | 1780 |