Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182496.2 | NP_872302.2 | 14 | 1550 | ||
XM_006719229.2 | XP_006719292.1 | 13 | 1343 | |||
XM_011537883.2 | XP_011536185.1 | 14 | 1580 | |||
XM_011537884.2 | XP_011536186.1 | 15 | 1693 | |||
XM_011537886.2 | XP_011536188.1 | 15 | 1741 | |||
XM_011537887.2 | XP_011536189.1 | 15 | 1778 | |||
XM_011537888.2 | XP_011536190.1 | 8 | 1664 | |||
GenBank mRNA | AK097408.1 | - | 14 | 1550 | ||
BC095479.1 | - | 14 | 1498 | 30389334 |