Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303133.1 | NP_001290062.1 | 3, 4 | 270 | ||
NM_152324.2 | NP_689537.1 | 3, 4 | 316 | |||
XM_017020387.1 | XP_016875876.1 | 3, 4 | 1624 | |||
XM_017020388.1 | XP_016875877.1 | 4, 5 | 478 | |||
GenBank mRNA | AW957098.1 | - | 3, 4 | 270 | ||
BC029889.1 | - | 3, 4 | 292 | 5170095 | ||
HY035354.1 | - | 3, 4 | 317 |