Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPACA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145248.4 | NP_660291.2 | 6 | 561 | ||
XM_011537466.1 | XP_011535768.1 | 7 | 736 | |||
XM_011537467.2 | XP_011535769.1 | 6 | 679 | |||
XM_011537468.2 | XP_011535770.1 | 8 | 797 | |||
XM_011537471.2 | XP_011535773.1 | 8 | 867 | |||
XM_011537472.2 | XP_011535774.1 | 5 | 548 | |||
XM_017020391.1 | XP_016875880.1 | 4 | 493 | |||
XM_017020392.1 | XP_016875881.1 | 6 | 614 | |||
GenBank mRNA | AK130866.1 | - | 6 | 561 | ||
BC016750.2 | - | 6 | 543 | 4837062 |