Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017923.1 | NP_001017923.1 | 5 | 1060 | ||
XM_011536408.1 | XP_011534710.1 | 4 | 361 | |||
GenBank mRNA | AF250395.1 | - | 5 | 1044 | ||
AK126985.1 | - | 3 | 1653 | |||
BC030258.1 | - | 4 | 798 | |||
BC105030.1 | - | 5 | 1053 | 8144033 | ||
BC105032.1 | - | 5 | 1053 | 8144035 |