Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152335.2 | NP_689548.2 | 5 | 582 | ||
XM_005254160.2 | XP_005254217.1 | 5 | 505 | |||
XM_017021915.1 | XP_016877404.1 | 7 | 798 | |||
XM_017021916.1 | XP_016877405.1 | 7 | 744 | |||
XM_017021917.1 | XP_016877406.1 | 7 | 730 | |||
GenBank mRNA | AK095509.1 | - | 5 | 474 | ||
AK308676.1 | - | 4 | 400 | |||
BX398159.2 | - | 4 | 498 | |||
DA090797.1 | - | 5 | 555 | |||
DA483185.1 | - | 5 | 474 | |||
DR007145.1 | - | 1 | 52 |