Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152339.3 | NP_689552.2 | 3 | 1576 | ||
XM_005256279.4 | XP_005256336.1 | 3 | 1993 | |||
GenBank mRNA | AF070574.1 | - | 1 | 807 | ||
AK127372.1 | - | 2 | 1380 | |||
BC039608.1 | - | 3 | 1554 | 5248678 | ||
BC053588.1 | - | 3 | 1558 | 5245683 |