Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152339.3 | NP_689552.2 | 3 | 2250 | ||
XM_005256279.4 | XP_005256336.1 | 3 | 2667 | |||
GenBank mRNA | AF070574.1 | - | 1 | 1481 | ||
AK127372.1 | - | 2 | 2054 | |||
BC027606.1 | - | 4 | 917 | |||
BC039608.1 | - | 3 | 2228 | 5248678 | ||
BC053588.1 | - | 3 | 2231 | 5245683 |