Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAQR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284511.1 | NP_001271440.1 | 3 | 1621 | ||
NM_001284512.1 | NP_001271441.1 | 2 | 1516 | |||
NM_001284513.1 | NP_001271442.1 | 3 | 1244 | |||
NM_001324118.1 | NP_001311047.1 | 4 | 1537 | |||
NM_152341.4 | NP_689554.2 | 3 | 1738 | |||
GenBank mRNA | AK054564.1 | - | 3 | 1322 | ||
AK092173.1 | - | 2 | 1203 | |||
AL832527.1 | - | 3 | 1351 | |||
AY424282.1 | - | 3 | 1308 | |||
BC033703.1 | - | 3 | 1396 | 4154019 |