Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242929.1 | NP_001229858.1 | 8 | 1067 | ||
NM_001308089.1 | NP_001295018.1 | 10 | 1257 | |||
NM_001324129.1 | NP_001311058.1 | 7 | 895 | |||
NM_001324130.1 | NP_001311059.1 | 10 | 1460 | |||
NM_133450.3 | NP_597707.1 | 11 | 1459 | |||
NR_040252.1 | 10 | 1462 | ||||
XM_005255119.3 | XP_005255176.1 | 9 | 1159 | |||
XM_005255121.3 | XP_005255178.1 | 7 | 857 | |||
XM_011522372.1 | XP_011520674.1 | 10 | 1389 | |||
XM_011522373.1 | XP_011520675.1 | 10 | 1271 | |||
XM_011522374.1 | XP_011520676.1 | 9 | 1005 | |||
XM_011522378.1 | XP_011520680.1 | 10 | 1127 | |||
XM_017022935.1 | XP_016878424.1 | 8 | 1067 | |||
XM_017022936.1 | XP_016878425.1 | 10 | 1380 | |||
GenBank mRNA | AB075857.1 | - | 8 | 1895 | ||
AK057329.1 | - | 7 | 1618 | |||
AK123371.1 | - | 7 | 856 | |||
AK123496.1 | - | 11 | 1460 | |||
AK301683.1 | - | 9 | 1087 | |||
BC050387.1 | - | 10 | 1098 | 5751728 | ||
BX333650.2 | - | 8 | 913 |