Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267576.1 | NP_001254505.1 | 9 | 1507 | ||
NM_153210.4 | NP_694942.3 | 9 | 1507 | |||
XM_011523639.2 | XP_011521941.1 | 8 | 1395 | |||
XM_011523640.1 | XP_011521942.1 | 8 | 1227 | |||
XM_011523641.1 | XP_011521943.1 | 9 | 952 | |||
XM_011523642.2 | XP_011521944.1 | 9 | 1552 | |||
XM_017024159.1 | XP_016879648.1 | 8 | 1448 | |||
XM_017024160.1 | XP_016879649.1 | 7 | 1434 | |||
XM_017024161.1 | XP_016879650.1 | 6 | 1520 | |||
XM_017024162.1 | XP_016879651.1 | 9 | 1552 | |||
GenBank mRNA | AJ583817.2 | - | 9 | 1414 | ||
BC136368.1 | - | 9 | 1507 | 9020355 | ||
BC144041.1 | - | 9 | 1507 | 9052561 | ||
BC171759.1 | - | 9 | 1507 | 9054412 | ||
BX648526.1 | - | 7 | 770 |