Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163673.1 | NP_001157145.1 | 10 | 2163 | ||
NM_001163809.1 | NP_001157281.1 | 10 | 5848 | |||
NM_001163811.1 | NP_001157283.1 | 10 | 2299 | |||
NM_152348.3 | NP_689561.2 | 11 | 2800 | |||
XM_011523651.2 | XP_011521953.1 | 11 | 2785 | |||
XM_017024184.1 | XP_016879673.1 | 10 | 5774 | |||
GenBank mRNA | AK074111.1 | - | 9 | 3423 | ||
AK074356.1 | - | 5 | 1390 | |||
AK091136.1 | - | 11 | 2784 | |||
AK122972.1 | - | 10 | 3110 | |||
AK123896.1 | - | 10 | 2290 | |||
AK127946.1 | - | 10 | 2299 | |||
AK298567.1 | - | 10 | 2163 | |||
AK300986.1 | - | 8 | 1780 | |||
AL834379.1 | - | 10 | 2624 | |||
BC092513.1 | - | 10 | 2575 | 3092839 | ||
BC114519.1 | - | 9 | 2058 | 40036610 | ||
BC114568.1 | - | 10 | 2125 | 40036619 |