Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INO80C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098817.1 | NP_001092287.1 | 7 | 729 | ||
NM_001308064.1 | NP_001294993.1 | 5 | 492 | |||
NM_194281.3 | NP_919257.2 | 5 | 621 | |||
XM_005258196.4 | XP_005258253.1 | 5 | 439 | |||
XM_005258197.4 | XP_005258254.1 | 5 | 652 | |||
XM_011525812.2 | XP_011524114.1 | 5 | 487 | |||
XM_017025549.1 | XP_016881038.1 | 5 | 459 | |||
GenBank mRNA | AK300660.1 | - | 7 | 681 | ||
BC039404.1 | - | 5 | 587 | 5300795 | ||
BC048989.1 | - | 6 | 549 | |||
BM758571.1 | - | 5 | 394 | |||
BQ441485.1 | - | 5 | 495 |