Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002836.3 | NP_001002836.2 | 3 | 1999 | ||
XM_011526445.2 | XP_011524747.1 | 3 | 3683 | |||
GenBank mRNA | AF000560.1 | - | 2 | 1794 | ||
BC012495.1 | - | 4 | 1291 | |||
BC077728.1 | - | 3 | 1901 | 6210543 |