Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMIGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169126.1 | NP_001162597.1 | 5 | 701 | ||
NM_001308232.1 | NP_001295161.1 | 4 | 353 | |||
NM_144615.2 | NP_653216.2 | 5 | 713 | |||
GenBank mRNA | AY358964.1 | - | 5 | 663 | ||
BC015655.1 | - | 5 | 713 | 4811204 | ||
HQ452865.1 | - | 4 | 307 | |||
HQ452866.1 | - | 2 | 151 |