Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPLAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300771.1 | NP_001287700.1 | 3 | 262 | ||
NM_001300772.1 | NP_001287701.1 | 3 | 180 | |||
NM_138794.4 | NP_620149.2 | 3 | 279 | |||
XM_011509182.2 | XP_011507484.1 | 4 | 290 | |||
XM_011509183.2 | XP_011507485.1 | 4 | 591 | |||
XM_017000271.1 | XP_016855760.1 | 4 | 290 | |||
GenBank mRNA | AA417842.1 | - | 3 | 178 | ||
AA424278.1 | - | 3 | 249 | |||
AK291542.1 | - | 3 | 233 | |||
AW967835.1 | - | 3 | 214 | |||
BC016711.1 | - | 3 | 233 | 4133970 | ||
BX282451.1 | - | 3 | 178 | |||
CB851909.1 | - | 3 | 269 | |||
DA822882.1 | - | 3 | 260 |