Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQGAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178229.4 | NP_839943.2 | 23 | 2731 | ||
XM_011509198.2 | XP_011507500.1 | 23 | 2720 | |||
XM_011509200.2 | XP_011507502.1 | 12 | 1900 | |||
XM_011509201.2 | XP_011507503.1 | 11 | 1289 | |||
XM_017000317.1 | XP_016855806.1 | 23 | 2717 | |||
XM_017000318.1 | XP_016855807.1 | 21 | 2474 | |||
GenBank mRNA | AB105103.1 | - | 23 | 2667 | ||
AK122653.1 | - | 9 | 939 | |||
AY253300.1 | - | 23 | 2656 | |||
BC131536.1 | - | 23 | 2689 | 40082932 |