Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178454.4 | NP_848549.3 | 6 | 682 | ||
XM_005270469.2 | XP_005270526.1 | 7 | 740 | |||
XM_005270470.1 | XP_005270527.1 | 7 | 863 | |||
XM_006710361.1 | XP_006710424.1 | 7 | 700 | |||
XM_006710362.1 | XP_006710425.1 | 7 | 673 | |||
XM_011540707.1 | XP_011539009.1 | 7 | 681 | |||
XM_011540708.1 | XP_011539010.1 | 7 | 761 | |||
XM_017000320.1 | XP_016855809.1 | 6 | 624 | |||
XM_017000321.1 | XP_016855810.1 | 6 | 483 | |||
GenBank mRNA | AK075350.1 | - | 2 | 128 | ||
AK309557.1 | - | 6 | 919 | |||
AL833098.1 | - | 6 | 1050 | |||
AY336747.1 | - | 6 | 560 | |||
AY358492.1 | - | 6 | 545 | |||
BC047025.1 | - | 5 | 497 | 5491665 | ||
BC091509.1 | - | 7 | 774 | 30513000 | ||
EF710624.1 | - | 4 | 425 |