Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080476.4 | NP_536724.1 | 3, 4 | 453 | ||
XM_017027664.1 | XP_016883153.1 | 3, 4 | 271 | |||
XM_017027665.1 | XP_016883154.1 | 3, 4 | 271 | |||
GenBank mRNA | AB086842.1 | - | 3, 4 | 259 | ||
AK075507.1 | - | 3, 4 | 278 | |||
AK301900.1 | - | 3, 4 | 253 | |||
AY358816.1 | - | 3, 4 | 283 | |||
AY422169.1 | - | 3, 4 | 453 | |||
BC030512.1 | - | 3, 4 | 264 | 4942673 |