Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C22orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173793.4 | NP_776154.3 | 3 | 1655 | ||
GenBank mRNA | AK095271.1 | - | 4 | 651 | ||
BC062599.1 | - | 3 | 1120 | 5248209 | ||
BX648073.1 | - | 1 | 424 |