Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TANGO2 SIRNA Assays ›
Gene Symbol:
TANGO2
Gene Name:
transport and golgi organization 2 homolog
Gene Aliases:
C22orf25, MECRCN
Chromosome Location:
Chr.22: 20017000 - 20067164 on Build GRCh38
UniGene ID:
Hs.474233
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s43378
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TANGO2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001283106.2
NP_001270035.1
9, 10
1072
NM_001283116.2
NP_001270045.1
8, 9
826
NM_001283129.2
NP_001270058.1
8, 9
1066
NM_001283148.2
NP_001270077.1
8, 9
824
NM_001283154.2
NP_001270083.1
8, 9
936
NM_001283179.2
NP_001270108.1
6, 7
767
NM_001283186.2
NP_001270115.1
6, 7
640
NM_001283199.2
NP_001270128.1
6, 7
713
NM_001283215.2
NP_001270144.1
6, 7
807
NM_001283235.2
NP_001270164.1
7, 8
711
NM_001283248.2
NP_001270177.1
5, 6
486
NM_001322141.1
NP_001309070.1
8, 9
1222
NM_001322142.1
NP_001309071.1
8, 9
982
NM_001322143.1
NP_001309072.1
8, 9
1139
NM_001322144.1
NP_001309073.1
8, 9
1064
NM_001322145.1
NP_001309074.1
6, 7
953
NM_001322146.1
NP_001309075.1
8, 9
1132
NM_001322147.1
NP_001309076.1
6, 7
880
NM_001322148.1
NP_001309077.1
8, 9
1143
NM_001322149.1
NP_001309078.1
6, 7
841
NM_001322150.1
NP_001309079.1
7, 8
1107
NM_001322153.1
NP_001309082.1
7, 8
867
NM_001322155.1
NP_001309084.1
7, 8
940
NM_001322160.1
NP_001309089.1
8, 9
1026
NM_001322163.1
NP_001309092.1
7, 8
886
NM_001322166.1
NP_001309095.1
6, 7
713
NM_001322167.1
NP_001309096.1
6, 7
765
NM_001322169.1
NP_001309098.1
6, 7
638
NM_001322171.1
NP_001309100.1
7, 8
784
NM_001322172.1
NP_001309101.1
7, 8
1076
NM_001322173.1
NP_001309102.1
7, 8
951
NM_001322174.1
NP_001309103.1
7, 8
838
NM_001322175.1
NP_001309104.1
7, 8
836
NM_152906.6
NP_690870.3
8, 9
938
NR_104274.2
6, 7
557
NR_104275.2
6, 7
797
NR_136206.1
6, 7
736
NR_136211.1
8, 9
922
NR_136212.1
7, 8
818
XM_011529863.1
XP_011528165.1
8, 9
975
XM_011529865.1
XP_011528167.1
6, 7
688
XM_011529867.1
XP_011528169.1
8, 9
921
XM_017028577.1
XP_016884066.1
8, 9
1055
XM_017028578.1
XP_016884067.1
8, 9
1074
XM_017028579.1
XP_016884068.1
8, 9
1103
XM_017028580.1
XP_016884069.1
9, 10
1192
XM_017028581.1
XP_016884070.1
8, 9
1186
XM_017028582.1
XP_016884071.1
9, 10
1269
XM_017028583.1
XP_016884072.1
8, 9
1016
XM_017028584.1
XP_016884073.1
8, 9
1279
XM_017028585.1
XP_016884074.1
6, 7
868
XM_017028586.1
XP_016884075.1
6, 7
793
XM_017028587.1
XP_016884076.1
8, 9
1030
XM_017028588.1
XP_016884077.1
7, 8
725
GenBank mRNA
AK092484.1
-
6, 7
595
AK289862.1
-
9, 10
1041
AK295210.1
-
7, 8
952
AK297075.1
-
5, 6
443
AK297922.1
-
8, 9
782
AK298593.1
-
7, 8
669
AK301054.1
-
6, 7
516
AK301366.1
-
6, 7
766
AK309398.1
-
5, 6
437
AK316020.1
-
8, 9
785
AK316056.1
-
8, 9
1046
AL713640.1
-
5, 6
566
BC041339.1
-
8, 9
901
5284126
BF726547.1
-
4, 5
423
BI911898.1
-
6, 7
662
BM048105.1
-
407
BU173566.1
-
8, 9
857
BX345246.2
-
6, 7
751
CF552364.1
-
7, 8
797
CR456355.1
-
7, 8
753
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0