Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2orf73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100396.1 | NP_001093866.1 | 5 | 749 | ||
XM_005264133.1 | XP_005264190.1 | 4 | 541 | |||
XM_005264134.3 | XP_005264191.1 | 4 | 624 | |||
XM_011532510.2 | XP_011530812.1 | 5 | 819 | |||
XM_011532513.2 | XP_011530815.1 | 4 | 722 | |||
XM_017003323.1 | XP_016858812.1 | 5 | 644 | |||
XM_017003324.1 | XP_016858813.1 | 4 | 707 | |||
GenBank mRNA | AK097617.1 | - | 4 | 541 | ||
BC031669.1 | - | 5 | 712 | |||
BC126244.1 | - | 4 | 510 | 8991960 | ||
BC126246.1 | - | 4 | 510 | 8991962 | ||
BC144191.1 | - | 3 | 394 | |||
BC144192.1 | - | 5 | 544 | 9052712 |