Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR9A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001656.1 | NP_001001656.1 | 1 | 69 | ||
GenBank mRNA | BC136819.1 | - | 1 | 128 | 9020810 | |
BC136821.1 | - | 1 | 128 | 9020812 |
Set Membership: |
G-protein coupled receptor |