Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301790.1 | NP_001288719.1 | 4 | 769 | ||
NM_152396.3 | NP_689609.2 | 5 | 904 | |||
XM_005264867.3 | XP_005264924.1 | 5 | 904 | |||
XM_005264870.4 | XP_005264927.1 | 6 | 1742 | |||
XM_006712970.3 | XP_006713033.1 | 5 | 780 | |||
XM_017005718.1 | XP_016861207.1 | 5 | 780 | |||
XM_017005719.1 | XP_016861208.1 | 4 | 762 | |||
XM_017005721.1 | XP_016861210.1 | 5 | 1600 | |||
GenBank mRNA | AK293370.1 | - | 4 | 762 | ||
BC022400.1 | - | 5 | 861 | 4693314 | ||
CX872865.1 | - | 2 | 273 |